Invitrogen
Manufacturer Code:PA521991
Catalog # PIPA521991
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 248 and 535 of SLC25A13 (Uniprot IDNo.Q9UJS0) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARALAR2 CITRIN CTLN2 calcium-binding mitochondrial carrier protein Aralar2 mitochondrial aspartate glutamate carrier 2 solute carrier family 25 member 13; Calcium-binding mitochondrial carrier protein Aralar2; Citrin; Mitochondrial aspartate glutamate carrier 2; solute carrier family 25 (aspartate/glutamate carrier), member 13; Solute carrier family 25 member 13; solute carrier family 25, member 13
Gene Aliases: AI785475; ARALAR2; CITRIN; CTLN2; Ctrn; SLC25A13
UniProt ID: (Human) O14575, (Mouse) Q9QXX4
Entrez Gene ID: (Human) 10165, (Mouse) 50799
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.