Invitrogen
Manufacturer Code:PA542430
Catalog # PIPA542430
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC25A12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: araceli hiperlarga; calcium binding mitochondrial carrier superfamily member Aralar1; Calcium-binding mitochondrial carrier protein Aralar1; Calcium-binding mitochondrial carrier protein Aralar1-like protein; Mitochondrial aspartate glutamate carrier 1; Solute carrier family 25 (aspartate/glutamate carrier) member 12 ARALAR ARALAR1 AGC1; solute carrier family 25 (aspartate/glutamate carrier), member 12; solute carrier family 25 (mitochondrial carrier, Aralar), member 12; Solute carrier family 25 member 12; solute carrier family 25, member 12
Gene Aliases: 2610002D09Rik; AGC1; AI839531; ARALAR; ARALAR1; B230107K20Rik; BB129864; RGD1561141; SLC25A12
UniProt ID: (Human) O75746, (Mouse) Q8BH59
Entrez Gene ID: (Horse) 100063768, (Pig) 100524283, (Dog) 478798, (Human) 8604, (Rabbit) 100350328, (Bovine) 539494, (Guinea pig) 100732340, (Mouse) 78830, (Rat) 362145
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.