Invitrogen
Manufacturer Code:PA596958
Catalog # PIPA596958
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-314 of human SLC25A11. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-oxoglutarate carrier; Mitochondrial 2-oxoglutarate/malate carrier protein; OGCP; solute carrier family 20 (oxoglutarate carrier), member 4; solute carrier family 25 (mitochondrial carrier; oxoglutarate carrier), member 11; Solute carrier family 25 member 11
Gene Aliases: 2310022P18Rik; 2oxoc; OGC; SLC20A4; SLC25A11
UniProt ID: (Human) Q02978, (Rat) P97700, (Mouse) Q9CR62
Entrez Gene ID: (Human) 8402, (Rat) 64201, (Mouse) 67863
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.