Invitrogen
Manufacturer Code:PA542428
Catalog # PIPA542428
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLC25A11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-oxoglutarate carrier; 2-oxoglutarate/malate carrier; Mitochondrial 2-oxoglutarate/malate carrier protein; OGCP; OMC; solute carrier family 20 (oxoglutarate carrier), member 4; solute carrier family 25 (mitochondrial carrier; oxoglutarate carrier), member 11; Solute carrier family 25 member 11
Gene Aliases: 2310022P18Rik; 2oxoc; fa07h09; OGC; SLC20A4; SLC25A11; wu:fa07h09; zgc:86898
UniProt ID: (Bovine) P22292, (Human) Q02978, (Rat) P97700, (Mouse) Q9CR62
Entrez Gene ID: (Bovine) 282523, (Sheep) 100147782, (Horse) 100061330, (Rabbit) 100342399, (Dog) 479470, (Human) 8402, (Rat) 64201, (Mouse) 67863, (Guinea pig) 100722970, (Zebrafish) 415189
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.