Invitrogen
Manufacturer Code:PA527510
Catalog # PIPA527510
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 92 and 314 of Human SLC25A11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mitochondrial 2-oxoglutarate/malate carrier protein; OGC SLC20A4 OGCP mitochondrial 2-oxoglutarate/malate carrier protein solute carrier family 20 (oxoglutarate carrier) member 4 solute carrier family 25 member 11; OGCP; solute carrier family 20 (oxoglutarate carrier), member 4; solute carrier family 25 (mitochondrial carrier; oxoglutarate carrier), member 11; Solute carrier family 25 member 11
Gene Aliases: OGC; SLC20A4; SLC25A11
UniProt ID: (Human) Q02978
Entrez Gene ID: (Human) 8402
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.