Novus Biologicals
Manufacturer Code:NBP2835430.1ML
Catalog # NB031472
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human SLC25A10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dicarboxylate ion carrier; dicarboxylate ion carrier DICSolute carrier family 25 member 10 member 10 mitochondrial dicarboxylate carrier solute carrier family 25 (mitochondrial carrier dicarboxylate transporter); Mitochondrial dicarboxylate carrier; solute carrier family 25 (mitochondrial carrier; dicarboxylate transporter), member 10; Solute carrier family 25 member 10
Gene Aliases: DIC; SLC25A10
UniProt ID: (Human) Q9UBX3
Entrez Gene ID: (Human) 1468
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.