Proteintech
Manufacturer Code:120861AP150UL
Catalog # 50-172-7701
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC25A10 Fusion Protein Ag2719 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DIC SLC25A10; dicarboxylate ion carrier; dicarboxylate transporter), member 10; Mitochondrial dicarboxylate carrier; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 10; solute carrier family 25 (mitochondrial carrier; dicarboxylate transporter), member 10; Solute carrier family 25 member 10
Gene Aliases: DIC; SLC25A10
UniProt ID: (Human) Q9UBX3, (Mouse) Q9QZD8
Entrez Gene ID: (Human) 1468, (Mouse) 27376, (Rat) 170943
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.