Invitrogen
Manufacturer Code:PA585163
Catalog # PIPA585163
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human SLC25A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Citrate transport protein; citrate transporter) member 1; CTP; CTP antibody SEA antibody SLC20A3 antibody SLC25A1 antibody citrate transport protein antibody solute carrier family 20 (mitochondrial citrate transporter) member 3 antibody tricarboxylate transport protein mitochondrial antibody tricarboxylate carrier protein antibody solute carrier family 25 (mitochondrial carrier citrate transporter) member 1 antibody; DiGeorge syndrome gene j; mitochondrial tricarboxylate carrier; solute carrier family 20 (mitochondrial citrate transporter), member 3; solute carrier family 25 (mitochondrial carrier; citrate transporter), member 1; Solute carrier family 25 member 1; Tricarboxylate carrier protein; Tricarboxylate transport protein, mitochondrial
Gene Aliases: 1300019P08Rik; 2610100G11Rik; AI194714; Cic; CTP; D2L2AD; Dgsj; SEA; SLC20A3; SLC25A1
UniProt ID: (Human) P53007, (Rat) P32089, (Mouse) Q8JZU2
Entrez Gene ID: (Human) 6576, (Rat) 29743, (Mouse) 13358
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.