Novus Biologicals
Manufacturer Code:NBP159599
Catalog # NBP159599
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLC25A1(solute carrier family 25 (mitochondrial carrier citrate transporter) member 1) The peptide sequence was selected from the middle region of SLC25A1. Peptide sequence NKPMNPLITGVFGAIAGAASVFGNTPLD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Citrate transport protein; Citrate transport protein CTPTricarboxylate carrier protein SEA SLC20A3member 3 solute carrier family 25 (mitochondrial carrier citrate transporter) member 1 Solute carrier family 25 member 1 tricarboxylate transport protein mitochondrial; CTP; solute carrier family 20 (mitochondrial citrate transporter), member 3; solute carrier family 25 (mitochondrial carrier; citrate transporter), member 1; Solute carrier family 25 member 1; Tricarboxylate carrier protein; Tricarboxylate transport protein, mitochondrial
Gene Aliases: CTP; D2L2AD; SEA; SLC20A3; SLC25A1
UniProt ID: (Human) P53007
Entrez Gene ID: (Human) 6576
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.