Proteintech
Manufacturer Code:152351AP150UL
Catalog # 50-554-534
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC25A1 Fusion Protein Ag7352 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CIC Citrate transport protein CTP SLC20A3 SLC25A1 Tricarboxylate carrier protein; Citrate transport protein; citrate transporter) member 1; CTP; DiGeorge syndrome gene j; mitochondrial tricarboxylate carrier; solute carrier family 20 (mitochondrial citrate transporter), member 3; solute carrier family 25 (mitochondrial carrier; citrate transporter), member 1; Solute carrier family 25 member 1; Tricarboxylate carrier protein; Tricarboxylate transport protein, mitochondrial
Gene Aliases: 1300019P08Rik; 2610100G11Rik; AI194714; Cic; CTP; D2L2AD; Dgsj; SEA; SLC20A3; SLC25A1
UniProt ID: (Human) P53007, (Rat) P32089, (Mouse) Q8JZU2
Entrez Gene ID: (Human) 6576, (Rat) 29743, (Mouse) 13358
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.