Invitrogen
Manufacturer Code:PA598527
Catalog # PIPA598527
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human Sodium/potassium/calcium exchanger 5 protein (240-298AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ion transporter JSX; Na(+)/K(+)/Ca(2+)-exchange protein 5; oculocutaneous albinism 6 (autosomal recessive); Sodium/potassium/calcium exchanger 5; solute carrier family 24 (sodium/potassium/calcium exchanger), member 5; Solute carrier family 24 member 5; solute carrier family 24, member 5
Gene Aliases: JSX; NCKX5; OCA6; SHEP4; SLC24A5
UniProt ID: (Human) Q71RS6
Entrez Gene ID: (Human) 283652
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.