Invitrogen
Manufacturer Code:PA567555
Catalog # PIPA567555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human SLC24A5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ion transporter JSX; Na(+)/K(+)/Ca(2+)-exchange protein 5; oculocutaneous albinism 6 (autosomal recessive); SLC24A5 solute carrier family 24member 5 JSX NCKX5 ion transporter JSX solute carrier family 24 (sodium/potassium/calcium exchanger)member 5; Sodium/potassium/calcium exchanger 5; solute carrier family 24 (sodium/potassium/calcium exchanger), member 5; Solute carrier family 24 member 5; solute carrier family 24, member 5
Gene Aliases: JSX; NCKX5; OCA6; SHEP4; SLC24A5
UniProt ID: (Human) Q71RS6
Entrez Gene ID: (Human) 283652
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.