Invitrogen
Manufacturer Code:PA518521
Catalog # PIPA518521
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (TTPDCRKKFWKN) corresponding to the internal amino acids of SLC24A5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: K-independent Na+/Ca2+ exchanger JSX; Na(+)/K(+)/Ca(2+)-exchange protein 5; Na-Ca exchanger 5; SLC24A5 solute carrier family 24member 5 JSX NCKX5 ion transporter JSX solute carrier family 24 (sodium/potassium/calcium exchanger)member 5; sodium/potassion/calcium exchanger 5; Sodium/potassium/calcium exchanger 5; Solute carrier family 24 member 5; solute carrier family 24, member 5
Gene Aliases: F630045L20Rik; JSX; NCKX5; NCX5; Oca6; Slc24a5
UniProt ID: (Mouse) Q8C261
Entrez Gene ID: (Mouse) 317750, (Rat) 311387
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.