Novus Biologicals
Manufacturer Code:NBP304861100UL
Catalog # NB096149
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 220-300 of human SLC24A5 (NP_995322.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ion transporter JSX; ion transporter JSX JSXsodium/potassium/calcium exchanger 5 Na(+)/K(+)/Ca(2+)-exchange protein 5 NCKX5solute carrier family 24 (sodium/potassium/calcium exchanger) member 5 SHEP4 Solute carrier family 24 member 5 solute carrier family 24 member 5; Na(+)/K(+)/Ca(2+)-exchange protein 5; oculocutaneous albinism 6 (autosomal recessive); Sodium/potassium/calcium exchanger 5; solute carrier family 24 (sodium/potassium/calcium exchanger), member 5; Solute carrier family 24 member 5; solute carrier family 24, member 5
Gene Aliases: JSX; NCKX5; OCA6; SHEP4; SLC24A5
UniProt ID: (Human) Q71RS6
Entrez Gene ID: (Human) 283652
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.