Abnova
Manufacturer Code:PAB7036
Catalog # 89-132-895
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human SLC24A5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of SLC24A5.
For Research Use Only
Protein Aliases: ion transporter JSX; JSX; Na(+)/K(+)/Ca(2+)-exchange protein 5; NCKX5; oculocutaneous albinism 6 (autosomal recessive); SHEP4; Sodium/potassium/calcium exchanger 5; solute carrier family 24 (sodium/potassium/calcium exchanger), member 5; Solute carrier family 24 member 5; solute carrier family 24, member 5
Gene Aliases: JSX; NCKX5; OCA6; SHEP4; SLC24A5
UniProt ID: (Human) Q71RS6
Entrez Gene ID: (Human) 283652
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.