Proteintech
Manufacturer Code:277471AP150UL
Catalog # 50-173-5746
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC24A5 Fusion Protein Ag24254 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ion transporter JSX; JSX NCKX5 SHEP4 SLC24A5; K-independent Na+/Ca2+ exchanger JSX; Na(+)/K(+)/Ca(2+)-exchange protein 5; Na-Ca exchanger 5; oculocutaneous albinism 6 (autosomal recessive); sodium/potassion/calcium exchanger 5; Sodium/potassium/calcium exchanger 5; solute carrier family 24 (sodium/potassium/calcium exchanger), member 5; Solute carrier family 24 member 5; solute carrier family 24, member 5
Gene Aliases: F630045L20Rik; JSX; NCKX5; NCX5; OCA6; SHEP4; SLC24A5
UniProt ID: (Human) Q71RS6, (Mouse) Q8C261
Entrez Gene ID: (Human) 283652, (Mouse) 317750, (Rat) 311387
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.