Invitrogen
Manufacturer Code:PA542518
Catalog # PIPA542518
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLC24A4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: K-dependent Na/Ca exchanger NCKX4; Na(+)/K(+)/Ca(2+)-exchange protein 4; Sodium/potassium/calcium exchanger 4; solute carrier family 24 (sodium/potassium/calcium exchanger), member 4; Solute carrier family 24 member 4
Gene Aliases: A930002M03Rik; AI2A5; AW492432; NCKX4; SHEP6; SLC24A2; SLC24A4
UniProt ID: (Human) Q8NFF2, (Mouse) Q8CGQ8
Entrez Gene ID: (Human) 123041, (Rabbit) 100348850, (Dog) 607455, (Bovine) 615739, (Horse) 100063742, (Rat) 314396, (Mouse) 238384
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.