Invitrogen
Manufacturer Code:PA561020
Catalog # PIPA561020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SLC24A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Na(+)/K(+)/Ca(2+)-exchange protein 3; NCKX3; sodium calcium exchanger; Sodium/potassium/calcium exchanger 3; solute carrier family 24 (sodium/potassium/calcium exchanger), member 3; Solute carrier family 24 member 3
Gene Aliases: NCKX3; SLC24A3
UniProt ID: (Human) Q9HC58
Entrez Gene ID: (Human) 57419
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.