Invitrogen
Manufacturer Code:PA537224
Catalog # PIPA537224
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues in human SLC24A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Na(+)/K(+)/Ca(2+)-exchange protein 2; potassium-dependent sodium-calcium exchanger; Retinal cone Na-Ca+K exchanger; Sodium/potassium/calcium exchanger 2; Sodium/potassium/calcium exchanger 2 Na(+)/K(+)/Ca(2+)-exchange protein 2 Retinal cone Na-Ca+K exchanger Solute carrier family 24 member 2 NCKX2; solute carrier family 24 (sodium/potassium/calcium exchanger), member 2; Solute carrier family 24 member 2; solute carrier family 24, member 2
Gene Aliases: 2810021B17Rik; 6330417K15Rik; AI847460; NCKX2; SLC24A2
UniProt ID: (Human) Q9UI40, (Rat) O54701
Entrez Gene ID: (Human) 25769, (Mouse) 76376, (Rat) 84550
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.