Novus Biologicals
Manufacturer Code:NB300600
Catalog # NB300600
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic Peptide: D(90) L N D K I R D Y T P Q P(102) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Na(+)/K(+)/Ca(2+)-exchange protein 2; Retinal cone Na-Ca+K exchanger; retinal cone Na-Ca+K exchanger solute carrier family 24 (sodium/potassium/calcium exchanger) member 2; Sodium/potassium/calcium exchanger 2; solute carrier family 24 (sodium/potassium/calcium exchanger), member 2; Solute carrier family 24 member 2
Gene Aliases: NCKX2; SLC24A2
UniProt ID: (Human) Q9UI40
Entrez Gene ID: (Human) 25769
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.