Abnova
Manufacturer Code:H00009187A01
Catalog # 89-106-025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 162 a.a. &bsim 268 a.a) partial recombinant protein with GST tag.,SLC24A1 (NP_004718 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant SLC24A1.
For Research Use Only
Protein Aliases: HsT17412; KIAA0702; Na(+)/K(+)/Ca(2+)-exchange protein 1; NCKX; NCKX1; retinal rod Na+/Ca+/K+ exchanger; Retinal rod Na-Ca+K exchanger; RODX; Sodium/potassium/calcium exchanger 1; solute carrier family 24 (sodium/potassium/calcium exchanger), member 1; Solute carrier family 24 member 1
Gene Aliases: CSNB1D; HsT17412; KIAA0702; NCKX; NCKX1; RODX; SLC24A1
UniProt ID: (Human) O60721
Entrez Gene ID: (Human) 9187
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.