Invitrogen
Manufacturer Code:PA542447
Catalog # PIPA542447
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC23A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hSVCT2; membrane transporter; mSVCT2; Na(+)/L-ascorbic acid transporter 2; Nucleobase transporter-like 1 protein; sodium-coupled ascorbic acid transporter 2; sodium-coupled ascorbic acid transporter SVCT2; Sodium-dependent vitamin C transporter 2; sodium-dependent vitamin C transporter-2; solute carrier family 23 (ascorbic acid transporter), member 2; Solute carrier family 23 (nucleobase transporters) member 2 KIAA0238 NBTL1 SLC23A1 SVCT2 YSPL2; solute carrier family 23 (nucleobase transporters), member 1; solute carrier family 23 (nucleobase transporters), member 2; Solute carrier family 23 member 2; SVCT-2; SVCT2; testicular secretory protein Li 48; Yolk sac permease-like molecule 2
Gene Aliases: AI844736; hSVCT2; KIAA0238; mKIAA0238; NBTL1; SLC23A1; SLC23A2; SVCT2; YSPL2; YSPL3
UniProt ID: (Human) Q9UGH3, (Mouse) Q9EPR4, (Rat) Q9WTW8
Entrez Gene ID: (Bovine) 783536, (Dog) 403490, (Human) 9962, (Rabbit) 100009466, (Horse) 100052186, (Guinea pig) 100714147, (Mouse) 54338, (Rat) 50622
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.