Invitrogen
Manufacturer Code:PA5113352
Catalog # PIPA5113352
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Solute carrier family 23 member 2 protein (1-90AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI844736 BOS_13058 hSVCT2 Kiaa0238 mKIAA0238 mSVCT2 Na(+)/L-ascorbic acid transporter 2 NBTL1 Nucleobase transporter-like 1 protein SLC23A1 Slc23a2 sodium-coupled ascorbic acid transporter 2 sodium-coupled ascorbic acid transporter SVCT2 Sodium-dependent vitamin C transporter 2 sodium-dependent vitamin C transporter-2 solute carrier family 23 (ascorbic acid transporter) member 2 solute carrier family 23 (nucleobase transporters) member 1 solute carrier family 23 (nucleobase transporters) member 2 Solute carrier family 23 member 2 SVCT2 SVCT-2 testicular secretory protein Li 48 Yolk sac permease-like molecule 2 Yspl2 YSPL3; hSVCT2; Na(+)/L-ascorbic acid transporter 2; Nucleobase transporter-like 1 protein; Sodium-dependent vitamin C transporter 2; sodium-dependent vitamin C transporter-2; solute carrier family 23 (ascorbic acid transporter), member 2; solute carrier family 23 (nucleobase transporters), member 1; solute carrier family 23 (nucleobase transporters), member 2; Solute carrier family 23 member 2; testicular secretory protein Li 48; Yolk sac permease-like molecule 2
Gene Aliases: hSVCT2; KIAA0238; NBTL1; SLC23A1; SLC23A2; SVCT2; YSPL2
UniProt ID: (Human) Q9UGH3
Entrez Gene ID: (Human) 9962
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.