Invitrogen
Manufacturer Code:PA5113474
Catalog # PIPA5113474
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Solute carrier family 23 member 1 protein (173-259AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hSVCT1; hSVCT1 Na(+)/L-ascorbic acid transporter 1 SLC23A1 SLC23A2 Sodium-dependent vitamin C transporter 1 sodium-dependent vitamin C transporter-1 solute carrier family 23 (ascorbic acid transporter) member 1 solute carrier family 23 (nucleobase transporters) member 1 solute carrier family 23 (nucleobase transporters) member 2 Solute carrier family 23 member 1 SVCT1 Yolk sac permease-like molecule 3 YSPL3; Na(+)/L-ascorbic acid transporter 1; Sodium-dependent vitamin C transporter 1; sodium-dependent vitamin C transporter-1; solute carrier family 23 (ascorbic acid transporter), member 1; solute carrier family 23 (nucleobase transporters), member 1; solute carrier family 23 (nucleobase transporters), member 2; Solute carrier family 23 member 1; Yolk sac permease-like molecule 3
Gene Aliases: D18Ucla2; SLC23A1; SLC23A2; SVCT1; YSPL3
UniProt ID: (Human) Q9UHI7, (Mouse) Q9Z2J0
Entrez Gene ID: (Human) 9963, (Mouse) 20522
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.