Novus Biologicals
Manufacturer Code:NBP192396
Catalog # NBP192396
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:KKEEGERLSLEELKLNLQKEISLAKAKYTASDLFRIPMLR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hOAT3; hOAT3 MGC24086 OAT3Organic anion transporter 3 solute carrier family 22 (organic anion transporter) member 8 solute carrier family 22 member 8; Organic anion transporter 3; solute carrier family 22 (organic anion transporter), member 8; Solute carrier family 22 member 8
Gene Aliases: OAT3; SLC22A8
UniProt ID: (Human) Q8TCC7
Entrez Gene ID: (Human) 9376
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.