Novus Biologicals
Manufacturer Code:NBP294604100UL
Catalog # NB023732
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SLC22A8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hOAT3; hOAT3 MGC24086 OAT3Organic anion transporter 3 solute carrier family 22 (organic anion transporter) member 8 solute carrier family 22 member 8; Organic anion transporter 3; solute carrier family 22 (organic anion transporter), member 8; Solute carrier family 22 member 8
Gene Aliases: OAT3; SLC22A8
UniProt ID: (Human) Q8TCC7
Entrez Gene ID: (Human) 9376
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.