Abnova
Manufacturer Code:PAB8457
Catalog # 89-133-675
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human SLC22A8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SLC22A8.
For Research Use Only
Protein Aliases: hOAT3; MGC24086; OAT3; Organic anion transporter 3; solute carrier family 22 (organic anion transporter), member 8; Solute carrier family 22 member 8
Gene Aliases: OAT3; SLC22A8
UniProt ID: (Human) Q8TCC7
Entrez Gene ID: (Human) 9376
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.