Invitrogen
Manufacturer Code:PA542139
Catalog # PIPA542139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLC22A7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hOAT2; liver-specific transporter; Novel liver transporter; OAT2; Organic anion transporter 2; rbOAT2; Solute carrier family 22 (organic anion transporter) member 7 MGC24091 MGC45202 NLT OAT2; solute carrier family 22 (organic anion transporter), member 7; Solute carrier family 22 member 7
Gene Aliases: NLT; OAT2; SLC22A7
UniProt ID: (Rabbit) Q3YAW7, (Human) Q9Y694, (Bovine) A6QLW8, (Rat) Q5RLM2
Entrez Gene ID: (Horse) 100055149, (Rabbit) 100009335, (Human) 10864, (Bovine) 407224, (Guinea pig) 100714866, (Rat) 89776
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.