Novus Biologicals
Manufacturer Code:NBP162660
Catalog # NBP162660
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLC22A7(solute carrier family 22 (organic anion transporter) member 7) The peptide sequence was selected from the N terminal of SLC22A7. Peptide sequence LPGAPANFSHQDVWLEAHLPREPDGTLSSCLRFAYPQALPNTTLGEER |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hOAT2; hOAT2 liver-specific transporter NLTMGC45202 Novel liver transporter OAT2MGC24091 Organic anion transporter 2 solute carrier family 22 (organic anion transporter) member 7 solute carrier family 22 member 7; liver-specific transporter; Novel liver transporter; Organic anion transporter 2; solute carrier family 22 (organic anion transporter), member 7; Solute carrier family 22 member 7
Gene Aliases: NLT; OAT2; SLC22A7
UniProt ID: (Human) Q9Y694
Entrez Gene ID: (Human) 10864
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.