Invitrogen
Manufacturer Code:PA542020
Catalog # PIPA542020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLC22A6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hOAT1; hPAHT; hROAT1; Kidney-specific transport protein; mNKT; mROAT1; Novel kidney transcript; Organic anion transporter 1; organic cationic transporter-like 1; PAH transporter; para-aminohippurate transporter; rbROAT1; Renal organic anion transporter 1; renal organic anion transporter 1 (rbOAT1); ROAT1; rROAT1; solute carrier family 22 (organic anion transporter), member 6; solute carrier family 22 (organic anion transporter), member 6, like; Solute carrier family 22 member 6; solute carrier family 22, member 6
Gene Aliases: HOAT1; mOat1; NKT; oat; OAT1; oat3; Orctl1; PAHT; ROAT1; SLC22A6; zgc:77073
UniProt ID: (Bovine) Q864Z3, (Rabbit) Q9TSY7, (Human) Q4U2R8, (Mouse) Q8VC69, (Rat) O35956
Entrez Gene ID: (Bovine) 407180, (Rabbit) 100008851, (Human) 9356, (Dog) 476050, (Horse) 100064220, (Guinea pig) 100727206, (Mouse) 18399, (Rat) 29509, (Zebrafish) 404609
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.