Invitrogen
Manufacturer Code:PA537389
Catalog # PIPA537389
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length human SLC22A5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDSP High-affinity sodium-dependent carnitine cotransporter OCTN2 OCTN2VT Organic cation/carnitine transporter 2 S22A5 Slc22a5 Solute carrier family 22 (organic cation/carnitine transporter) member 5 Solute carrier family 22 member 5; CT1; high-affinity carnitine transporter; high-affinity sodium dependent carnitine cotransporter; High-affinity sodium-dependent carnitine cotransporter; integral membrane transport protein; juvenile visceral steatosis; Organic cation/carnitine transporter 2; solute carrier family 22 (organic cation transporter), member 5; Solute carrier family 22 member 5; solute carrier family 22, member 5; UST2r
Gene Aliases: CDSP; CT1; jvs; Lstpl; OCTN2; SLC22A5; UST2r
UniProt ID: (Human) O76082, (Mouse) Q9Z0E8, (Rat) O70594
Entrez Gene ID: (Human) 6584, (Mouse) 20520, (Rat) 29726
Molecular Function:
carbohydrate transporter
cation transporter
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.