Invitrogen
Manufacturer Code:PA5110381
Catalog # PIPA5110381
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human SLC22A5 (NP_003051.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDSP CT1 High-affinity sodium-dependent carnitine cotransporter jvs Lstpl Octn2 OCTN2VT Organic cation/carnitine transporter 2 S22A5 Slc22a5 Solute carrier family 22 member 5 UST2r; high-affinity sodium dependent carnitine cotransporter; High-affinity sodium-dependent carnitine cotransporter; juvenile visceral steatosis; Organic cation/carnitine transporter 2; Solute carrier family 22 member 5
Gene Aliases: CDSP; jvs; Lstpl; OCTN2; SLC22A5
UniProt ID: (Human) O76082, (Mouse) Q9Z0E8
Entrez Gene ID: (Human) 6584, (Mouse) 20520
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.