Invitrogen
Manufacturer Code:PA575776
Catalog # PIPA575776
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human SLC22A4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ergothioneine transporter; ET transporter; integral membrane transport protein; LSTP-Like 2; organic cation transporter 1; organic cation transporter OCTN1; Organic cation/carnitine transporter 1; solute carrier family (organic cation transporter), member 4; solute carrier family 22 (organic cation transporter), member 4; solute carrier family 22 (organic cation/ergothioneine transporter), member 4; solute carrier family 22 (organic cation/zwitterion transporter), member 4; Solute carrier family 22 member 4; Solute carrier family 22 member 4 Ergothioneine transporter ET transporter Organic cation/carnitine transporter 1 SLC22A4 ETT OCTN1 UT2H
Gene Aliases: ETT; OCTN1; SLC22A4; UT2H
UniProt ID: (Human) Q9H015, (Rat) Q9R141, (Mouse) Q9Z306
Entrez Gene ID: (Human) 6583, (Rat) 64037, (Mouse) 30805
Molecular Function:
carbohydrate transporter
cation transporter
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.