Abnova
Manufacturer Code:H00006583K
Catalog # 89-202-361
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SLC22A4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SLC22A4 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Ergothioneine transporter; ET transporter; integral membrane transport protein; MGC34546 MGC40524 OCTN1; Organic cation/carnitine transporter 1; solute carrier family 22 (organic cation/ergothioneine transporter), member 4; solute carrier family 22 (organic cation/zwitterion transporter), member 4; Solute carrier family 22 member 4
Gene Aliases: ETT; OCTN1; SLC22A4; UT2H
UniProt ID: (Human) Q9H015
Entrez Gene ID: (Human) 6583
Molecular Function:
carbohydrate transporter
cation transporter
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.