Invitrogen
Manufacturer Code:PA5111769
Catalog # PIPA5111769
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide corresponding to amino acid residues 83-96 of rat OCT3 (Extracellular 1 st loop) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-Oct EMT EMTH Extraneuronal monoamine transporter OCT3 Orct3 Organic cation transporter 3 Slc22a3 Slca22a3 Solute carrier family 22 member 3; Organic cation transporter 3; solute carrier family 22 (extraneuronal monoamine transporter), member 3; Solute carrier family 22 member 3; solute carrier family 22, member 3
Gene Aliases: EMT; Oct3; Orct3; Slc22a3; Slca22a3
UniProt ID: (Rat) O88446, (Mouse) Q9WTW5
Entrez Gene ID: (Rat) 29504, (Mouse) 20519
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.