Invitrogen
Manufacturer Code:MA524289
Catalog # PIMA524289
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 640438 |
Immunogen | NS0 mouse myeloma cell line transfected with human SLC-22A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: hOCT2; Organic cation transporter 2; solute carrier family 22 (organic cation transporter), member 2; Solute carrier family 22 member 2; Solute carrier family 22 member 2 Organic cation transporter 2 hOCT2 OCT2
Gene Aliases: OCT2; SLC22A2
UniProt ID: (Human) O15244
Entrez Gene ID: (Human) 6582
Molecular Function:
carbohydrate transporter
cation transporter
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.