Invitrogen
Manufacturer Code:PA589602
Catalog # PIPA589602
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 44-140 of human SLC22A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Organic cation transporter 2; Solute carrier family 22 member 2; solute carrier family 22, member 2
Gene Aliases: Oct2; Orct2; Slc22a2
UniProt ID: (Mouse) O70577
Entrez Gene ID: (Mouse) 20518
Molecular Function:
carbohydrate transporter
cation transporter
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.