Invitrogen
Manufacturer Code:PA5112715
Catalog # PIPA5112715
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Solute carrier family 22 member 2 protein (48-135AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-Oct hOCT2 Oct2 OCT2r Orct2 Organic cation transporter 2 rOCT2 Slc22a2 solute carrier family 22 (organic cation transporter) member 2 Solute carrier family 22 member 2 solute carrier family 22 member 2; hOCT2; Organic cation transporter 2; rOCT2; solute carrier family 22 (organic cation transporter), member 2; Solute carrier family 22 member 2; solute carrier family 22, member 2
Gene Aliases: OCT2; OCT2r; Orct2; rOCT2; SLC22A2
UniProt ID: (Human) O15244, (Rat) Q9R0W2, (Mouse) O70577
Entrez Gene ID: (Human) 6582, (Rat) 29503, (Mouse) 20518
Molecular Function:
carbohydrate transporter
cation transporter
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.