Proteintech
Manufacturer Code:135941AP150UL
Catalog # 50-172-8861
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC22A2 Fusion Protein Ag4500 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hOCT2; hOCT2 OCT2 Organic cation transporter 2 SLC22A2; Organic cation transporter 2; rOCT2; solute carrier family 22 (organic cation transporter), member 2; Solute carrier family 22 member 2; solute carrier family 22, member 2
Gene Aliases: OCT2; OCT2r; Orct2; rOCT2; SLC22A2
UniProt ID: (Human) O15244, (Rat) Q9R0W2, (Mouse) O70577
Entrez Gene ID: (Human) 6582, (Rat) 29503, (Mouse) 20518
Molecular Function:
carbohydrate transporter
cation transporter
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.