Proteintech
Manufacturer Code:116462AP150UL
Catalog # 50-172-7340
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC22A18 Fusion Protein Ag2245 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beckwith-Wiedemann syndrome chromosomal region 1 candidate gene A protein; BWR1A BWSCR1A HET IMPT1 ITM ORCTL 2 ORCTL2 p45 BWR1A SLC22A18 SLC22A1L TSSC5; Efflux transporter-like protein; imprinted multi-membrane spanning polyspecific transporter-related protein 1; Imprinted multi-membrane-spanning polyspecific transporter-related protein 1; Multi-membrane-spanning polyspecific transporter; ORCTL-2; Organic cation transporter-like protein 2; organic cationic transporter-like 2; p45 Beckwith-Wiedemann region 1A; p45-Beckwith-Wiedemann region 1 A; p45-BWR1A; solute carrier family 22 (organic cation transporter), member 18; Solute carrier family 22 member 1-like; Solute carrier family 22 member 18; solute carrier family 22, member 18; tumor suppressing subtransferable candidate 5; Tumor-suppressing STF cDNA 5 protein; Tumor-suppressing subchromosomal transferable fragment candidate gene 5 protein
Gene Aliases: AW260131; BWR1A; BWSCR1A; HET; IMPT1; ITM; ORCTL2; p45-BWR1A; SLC22A18; SLC22A1L; TSSC5
UniProt ID: (Human) O60680, (Mouse) Q78KK3, (Rat) Q6AY78
Entrez Gene ID: (Human) 5002, (Mouse) 18400, (Rat) 309131
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.