Novus Biologicals
Manufacturer Code:NBP159889
Catalog # NBP159889
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLC22A17(solute carrier family 22 member 17) The peptide sequence was selected from the middle region of SLC22A17. Peptide sequence HCYQPVGGGGSPSDFYLCSLLASGTAALACVFLGVTVDRFGRRGILLLSM. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 24p3 receptor; 24p3R; 24p3R BOCTNGALR3 BOIThBOIT Brain-type organic cation transporter Lipocalin-2 receptor Neutrophil gelatinase-associated lipocalin receptor NgalR NGALR2 NGALRNGAL receptor potent brain type organic ion transporter solute carrier family 22 (organic cation transporter) member 17 solute carrier family 22 member 1724p3 receptor solute carrier family 22 member 17; Brain-type organic cation transporter; Lipocalin-2 receptor; Neutrophil gelatinase-associated lipocalin receptor; NGAL receptor; NgalR; potent brain type organic ion transporter; solute carrier family 22 (organic cation transporter), member 17; Solute carrier family 22 member 17; solute carrier family 22, member 17
Gene Aliases: 24p3R; BOCT; BOIT; hBOIT; NGALR; NGALR2; NGALR3; SLC22A17
UniProt ID: (Human) Q8WUG5
Entrez Gene ID: (Human) 51310
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.