Novus Biologicals
Manufacturer Code:NBP120975
Catalog # NBP120975
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-PETKRKLLPEVLRD corresponding to internal region according to NP_057693.3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 24p3 receptor; 24p3R; 24p3R BOCTNGALR3 BOIThBOIT Brain-type organic cation transporter Lipocalin-2 receptor Neutrophil gelatinase-associated lipocalin receptor NgalR NGALR2 NGALRNGAL receptor potent brain type organic ion transporter solute carrier family 22 (organic cation transporter) member 17 solute carrier family 22 member 1724p3 receptor solute carrier family 22 member 17; Brain-type organic cation transporter; Lipocalin-2 receptor; Neutrophil gelatinase-associated lipocalin receptor; NGAL receptor; NgalR; potent brain type organic ion transporter; solute carrier family 22 (organic cation transporter), member 17; Solute carrier family 22 member 17; solute carrier family 22, member 17
Gene Aliases: 24p3R; BOCT; BOIT; hBOIT; NGALR; NGALR2; NGALR3; SLC22A17
UniProt ID: (Human) Q8WUG5
Entrez Gene ID: (Human) 51310
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.