Abnova
Manufacturer Code:PAB7134
Catalog # 89-132-981
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human SLC22A16. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of SLC22A16.
For Research Use Only
Protein Aliases: Carnitine transporter 2; CT2; dJ261K5.1; epididymis secretory protein Li 18; flipt 2; FLIPT2; fly-like putative organic ion transporter 2; Fly-like putative transporter 2; OCT6; OKB1; organic cation transporter 6; Organic cation transporter OKB1; Organic cation/carnitine transporter 6; solute carrier family 22 (organic cation transporter), member 16; solute carrier family 22 (organic cation/carnitine transporter), member 16; Solute carrier family 22 member 16; WUGSC:RG331P03.1
Gene Aliases: CT2; dJ261K5.1; FLIPT2; HEL-S-18; OAT6; OCT6; OKB1; SLC22A16
UniProt ID: (Human) Q86VW1
Entrez Gene ID: (Human) 85413
Molecular Function:
carbohydrate transporter
cation transporter
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.