Invitrogen
Manufacturer Code:PA569275
Catalog # PIPA569275
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of human SLC22A15 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Flipt 1; fly-like putative organic ion transporter 1; Fly-like putative transporter 1; solute carrier family 22 (organic cation transporter), member 15; Solute carrier family 22 member 15; solute carrier family 22, member 15; trans-like protein
Gene Aliases: FLIPT1; PRO34686; SLC22A15; UNQ9429/PRO34686
UniProt ID: (Human) Q8IZD6
Entrez Gene ID: (Bovine) 512417, (Human) 55356, (Dog) 483136, (Horse) 100065881, (Rabbit) 100341700, (Rat) 310732
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.