Invitrogen
Manufacturer Code:PA554080
Catalog # PIPA554080
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SLC22A15 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Flipt 1; fly-like putative organic ion transporter 1; Fly-like putative transporter 1; solute carrier family 22 (organic cation transporter), member 15; Solute carrier family 22 member 15; solute carrier family 22, member 15; trans-like protein
Gene Aliases: FLIPT1; PRO34686; SLC22A15; UNQ9429/PRO34686
UniProt ID: (Human) Q8IZD6
Entrez Gene ID: (Human) 55356
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.