Invitrogen
Manufacturer Code:PA542438
Catalog # PIPA542438
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLC22A13 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ORCTL-3; Organic cation transporter-like 3; organic cationic transporter-like 3; organic-cation transporter like 3; Solute carrier family 22 (organic anion transporter) member 13 OCTL1 OCTL3 ORCTL3 OAT10 ORCTL-3; solute carrier family 22 (organic anion transporter), member 13; solute carrier family 22 (organic anion/urate transporter), member 13; solute carrier family 22 (organic cation transporter), member 13; Solute carrier family 22 member 13; solute carrier family 22, member 13
Gene Aliases: AI648912; OAT10; OCTL1; OCTL3; ORCTL-3; ORCTL3; SLC22A13
UniProt ID: (Human) Q9Y226, (Mouse) Q6A4L0
Entrez Gene ID: (Dog) 485587, (Human) 9390, (Horse) 100053841, (Pig) 100518094, (Mouse) 102570, (Rat) 316062
Molecular Function:
carbohydrate transporter
cation transporter
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.