Abnova
Manufacturer Code:H00009390A01
Catalog # 89-106-071
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 38 a.a. &bsim 139 a.a) partial recombinant protein with GST tag.,SLC22A13 (NP_004247 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant SLC22A13.
For Research Use Only
Protein Aliases: OAT10; OCTL1; OCTL3; ORCTL-3; ORCTL3; Organic cation transporter-like 3; organic cationic transporter-like 3; organic-cation transporter like 3; solute carrier family 22 (organic anion transporter), member 13; solute carrier family 22 (organic anion/urate transporter), member 13; Solute carrier family 22 member 13; solute carrier family 22, member 13
Gene Aliases: OAT10; OCTL1; OCTL3; ORCTL-3; ORCTL3; SLC22A13
UniProt ID: (Human) Q9Y226
Entrez Gene ID: (Human) 9390
Molecular Function:
carbohydrate transporter
cation transporter
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.