Invitrogen
Manufacturer Code:PA542510
Catalog # PIPA542510
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLC22A12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Organic anion transporter 4-like protein; Renal-specific transporter; RST; solute carrier family 22 (organic anion/cation transporter), member 12; Solute carrier family 22 (organic anion/urate transporter) member 12 OAT4L RST URAT1; solute carrier family 22 (organic anion/urate transporter), member 12; Solute carrier family 22 member 12; Urate anion exchanger 1; urate transporter 1
Gene Aliases: OAT4L; OATL4; RST; SLC22A12; UNQ6453/PRO34004; URAT1
UniProt ID: (Human) Q96S37
Entrez Gene ID: (Human) 116085
Molecular Function:
carbohydrate transporter
cation transporter
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.