Invitrogen
Manufacturer Code:PA577387
Catalog # PIPA577387
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)ENLGRRKSKAKENT corresponding to amino acid residues 530- 543 of rat Organic Cation Transporter 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mOCT1; Organic Cation Transporter 1; rOCT1; Solute carrier family 22 member 1; solute carrier family 22, member 1
Gene Aliases: Lx1; Oct1; Orct; Orct1; Roct1; Slc22a1
UniProt ID: (Rat) Q63089, (Mouse) O08966
Entrez Gene ID: (Rat) 24904, (Mouse) 20517
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.