Invitrogen
Manufacturer Code:PA576878
Catalog # PIPA576878
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human SLC22A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hOCT1; mOCT1; Organic cation transporter 1; rOCT1; solute carrier family 22 (organic cation transporter), member 1; Solute carrier family 22 member 1; Solute carrier family 22 member 1 Organic cation transporter 1 hOCT1 SLC22A1 OCT1; solute carrier family 22, member 1
Gene Aliases: HOCT1; Lx1; OCT1; oct1_cds; Orct; Orct1; Roct1; SLC22A1
UniProt ID: (Human) O15245, (Rat) Q63089, (Mouse) O08966
Entrez Gene ID: (Human) 6580, (Rat) 24904, (Mouse) 20517
Molecular Function:
carbohydrate transporter
cation transporter
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.